Learn More
Cayman Chemical 9-OxoDE-d3 25ug
SDP
Catalog No. 502947875
Encompass_Preferred
An internal standard for the quantification of 9-OxoODE by GC- or LC-MS
Catalog No. 50-294-7875 Supplier Cayman Chemical Supplier No. 33842025 UG
An internal standard for the quantification of 9-OxoODE by GC- or LC-MS