Learn More
Cayman Chemical 9-Oxononanoic Acid, Purity: ≥98%, CAS: 2553-17-5, 10 mg
SDP
Catalog No. 502988203
Encompass_Preferred
An oxidized fatty acid; formed via the autoxidation of linoleic acid; increases PLA2 activity and production of TXB2 in isolated human plasma; decreases hepatic de novo fatty acid synthesis and increases hepatic carnitine palmitoyltransferase activity in rats
Catalog No. 50-298-8203 Supplier Cayman Chemical Supplier No. 2988210 MG
An oxidized fatty acid; formed via the autoxidation of linoleic acid; increases PLA2 activity and production of TXB2 in isolated human plasma; decreases hepatic de novo fatty acid synthesis and increases hepatic carnitine palmitoyltransferase activity in rats