Learn More
Medchemexpress LLC 9-oxooctadecanoic acid | 4114-74-3 | MFCD05864058 | >98.0% | 298.46 g·mol-1 | C18H34O3 | 10 MG
SDP
Catalog No. 5000283923
Encompass_Preferred
9-Oxooctadecanoic acid is an oxidized C18 fatty acid supplied as a research-grade biochemical reagent and analytical reference standard for studies of lipid oxidation and fatty acid metabolism. It is provided in small package sizes for laboratory use and is suitable for analytical methods such as HPLC and mass spectrometry.
  • High purity (>98%) suitable for analytical and biochemical applications.
  • Commonly used as a reference standard in lipid analysis workflows.
  • Compatible with HPLC and mass spectrometry assays.
  • Supplied in small research quantities (for example, 10 mg) for analytical workflows.
  • Colorless to light yellow appearance for visual inspection of material.
Catalog No. 50-002-83923 Supplier Medchemexpress LLC Supplier No. HYW67004410MG
9-Oxooctadecanoic acid is an oxidized C18 fatty acid supplied as a research-grade biochemical reagent and analytical reference standard for studies of lipid oxidation and fatty acid metabolism. It is provided in small package sizes for laboratory use and is suitable for analytical methods such as HPLC and mass spectrometry.
  • High purity (>98%) suitable for analytical and biochemical applications.
  • Commonly used as a reference standard in lipid analysis workflows.
  • Compatible with HPLC and mass spectrometry assays.
  • Supplied in small research quantities (for example, 10 mg) for analytical workflows.
  • Colorless to light yellow appearance for visual inspection of material.