Learn More
Cayman Chemical 9-PAHSA 13C4 250ug
SDP
Catalog No. 502968485
Encompass_Preferred
An internal standard for the quantification of 9-PAHSA by NMR or GC- or LC-MS
This item is not returnable. View return policy
Catalog No. 50-296-8485 Supplier Cayman Chemical Supplier No. 17725250 UG
An internal standard for the quantification of 9-PAHSA by NMR or GC- or LC-MS