Learn More
Cayman Chemical 9-PAHSA-d31 100ug
SDP
Catalog No. 502979542
Encompass_Preferred
An internal standard for the quantification of 9-PAHSA by GC- or LC-MS
Catalog No. 50-297-9542 Supplier Cayman Chemical Supplier No. 17397100 UG
An internal standard for the quantification of 9-PAHSA by GC- or LC-MS