Learn More
Andwin Scientific 9-PANEL DRUG CARD TEST PK/25
SDP
Catalog No. 50465174
Encompass_Preferred
The 9-panel drug test card is used to quickly test urine in order to uncover the use of drugs, such as cocaine, amphetamine, methamphetamine, marijuana, opiate 2000, phencyclidine, barbiturate, and bensodiazepine. This product is sold in a pack of 25.
Catalog No. 50-465-174 Supplier Andwin Scientific Supplier No. 709420
The 9-panel drug test card is used to quickly test urine in order to uncover the use of drugs, such as cocaine, amphetamine, methamphetamine, marijuana, opiate 2000, phencyclidine, barbiturate, and bensodiazepine. This product is sold in a pack of 25.