Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Phenanthreneboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. P10935G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1093-5G 5 g
P10931G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P1093-5G Supplier TCI America Supplier No. P10935G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 68572-87-2
Molecular Formula C14H11BO2
Molecular Weight (g/mol) 222.05
MDL Number MFCD00143524
InChI Key JCDAUYWOHOLVMH-UHFFFAOYSA-N
PubChem CID 11775704

Specifications

Quantity 5 g
Synonym 9-phenanthreneboronic acid, 9-phenanthracenylboronic acid, phenanthrene-9-boronic acid, 9-phenanthrenylboronic acid, 9-phenanthrene boronic acid
SMILES B(C1=CC2=CC=CC=C2C3=CC=CC=C13)(O)O
IUPAC Name phenanthren-9-ylboronic acid
Formula Weight 222.05
Physical Form Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.