Learn More
TARGETMOL CHEMICALS INC 9-Phenanthrol 200MG
SDP
Catalog No. 502876322
Encompass_Preferred
Also available in 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. 9-Phenanthrol is inhibitor of the transient receptor potential melastatin 4 (TRPM) channel, a Ca2+ -activated non-selective cation channel. Purity 98.72%
Catalog No. 50-287-6322 Supplier TARGETMOL CHEMICALS INC Supplier No. T22537200MG
Also available in 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. 9-Phenanthrol is inhibitor of the transient receptor potential melastatin 4 (TRPM) channel, a Ca2+ -activated non-selective cation channel. Purity 98.72%