Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Phenylxanthen-9-ol, TCI America™
SDP

Catalog No. P12695G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P12695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P12695G Supplier TCI America Supplier No. P12695G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 596-38-3
Molecular Formula C19H14O2
Molecular Weight (g/mol) 274.319
MDL Number MFCD00005058
InChI Key CVZUPKFPOSRRSK-UHFFFAOYSA-N
Synonym 9-phenyl-9h-xanthen-9-ol, 9h-xanthen-9-ol, 9-phenyl, 9-phenyl-9-xanthenol, 9-phenylxanthydrol, 9-phenyl-xanthenol, acmc-209meu, 9-phenyl-9h-xanthen-9-ol #
PubChem CID 68997
IUPAC Name 9-phenylxanthen-9-ol
SMILES C1=CC=C(C=C1)C2(C3=CC=CC=C3OC4=CC=CC=C42)O

Specifications

Melting Point 162°C
Color White
Quantity 5 g
Formula Weight 274.32
Physical Form Crystalline Powder
Chemical Name or Material 9-Phenylxanthen-9-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.