Learn More
Medicus Health 9 Place Test Tube Rack
SDP
Catalog No. NC3790344
Encompass
• 9 Place Test Tube Rack for Phlebotomy Trays
• 9 Place racks fit perfectly into 2637M1 Phlebotomy Trays to secure tubes
• Holds 8-13mm Dia. Tubes
• Dimensions: 3" L x 3" W x 3" H
Catalog No. NC3790344 Supplier Medicus Health Supplier No. 2637M2
• 9 Place Test Tube Rack for Phlebotomy Trays
• 9 Place racks fit perfectly into 2637M1 Phlebotomy Trays to secure tubes
• Holds 8-13mm Dia. Tubes
• Dimensions: 3" L x 3" W x 3" H