Learn More
Chemglass Life Sciences Stirrer, Magnetic, Nine Position, Dura-Mag, 120V, with Factory Installed Alarm
SDP
Catalog No. NC1752641
Encompass
The Chemglass Life Sciences Nine-Position magnetic stirrer is a rugged design for 24/7 laboratory or production applications. The small footprint is ideal for CO² incubators with maximum capacity of (9) 500mL spinner vessels. Optional factory installed motion alarm available.
Catalog No. NC1752641 Supplier Chemglass Life Sciences Supplier No. CLS4100A9
The Chemglass Life Sciences Nine-Position magnetic stirrer is a rugged design for 24/7 laboratory or production applications. The small footprint is ideal for CO² incubators with maximum capacity of (9) 500mL spinner vessels. Optional factory installed motion alarm available.