Learn More
Medchemexpress LLC HY-100079 1mg Medchemexpress, 9-Propenyladenine CAS:1446486-33-4 Purity:>98%
SDP
Catalog No. 501931711
Encompass_Preferred
Medchemexpress, HY-100079 1mg 9-Propenyladenine CAS:1446486-33-4 9-Propenyladenine is a mutagenic impurity in tenofovir disoproxil fumarate. Tenofovir is an antiretroviral drug known as nucleotide analogue reverse transcriptase inhibitors, which block reverse transcriptase, a crucial virus enzyme in HIV-1 and HBV. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1711 Supplier Medchemexpress LLC Supplier No. HY1000791MG
Add to Cart
Add to Cart
Medchemexpress, HY-100079 1mg 9-Propenyladenine CAS:1446486-33-4 9-Propenyladenine is a mutagenic impurity in tenofovir disoproxil fumarate. Tenofovir is an antiretroviral drug known as nucleotide analogue reverse transcriptase inhibitors, which block reverse transcriptase, a crucial virus enzyme in HIV-1 and HBV. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.