Learn More
Cayman Chemical 9 RHODE cholesTRyl esTR 100ug
SDP
Catalog No. 502917843
Encompass_Preferred
9(R)-HODE cholesteryl ester was originally extracted from atherosclerotic lesions.{2227} It remains uncertain whether the oxidized fatty acid portion of the molecule results from enzymatic lipoxygenation or from random lipid peroxidation.{1126} 9(R)-HODE cholesteryl ester can be used as a standard for analysis of chiral HODE cholesteryl esters.
Catalog No. 50-291-7843 Supplier Cayman Chemical Supplier No. 38406100 UG
9(R)-HODE cholesteryl ester was originally extracted from atherosclerotic lesions.{2227} It remains uncertain whether the oxidized fatty acid portion of the molecule results from enzymatic lipoxygenation or from random lipid peroxidation.{1126} 9(R)-HODE cholesteryl ester can be used as a standard for analysis of chiral HODE cholesteryl esters.