Learn More
Cayman Chemical 9 S 12 S 13 S-TrIHODE 100ug
SDP
Catalog No. 502916041
Encompass_Preferred
An oxylipin; tumor levels are decreased compared with adjacent healthy tissue in dogs with mammary cancer
Catalog No. 50-291-6041 Supplier Cayman Chemical Supplier No. 38950100 UG
An oxylipin; tumor levels are decreased compared with adjacent healthy tissue in dogs with mammary cancer