Learn More
Cayman Chemical 9 S-HODE-13C18 100ug
SDP
Catalog No. 502989537
Encompass_Preferred
An internal standard for the quantification of 9-HODE by GC- or LC-MS
Catalog No. 50-298-9537 Supplier Cayman Chemical Supplier No. 31160100 UG
An internal standard for the quantification of 9-HODE by GC- or LC-MS