Learn More
Cayman Chemical 9 S-HODE-d4 50ug
SDP
Catalog No. 502947836
Encompass_Preferred
An internal standard for the quantification of 9-HODE by GC- or LC-MS
Catalog No. 50-294-7836 Supplier Cayman Chemical Supplier No. 33841050 UG
An internal standard for the quantification of 9-HODE by GC- or LC-MS