Learn More
Chemglass Life Sciences #9 Outer Joint, 11mm X 1.5W Tubing
SDP
Catalog No. 501531916
Encompass_Preferred
#9 Outer Joint, 11mm X 1.5W Tubing. Standard taper flask length outer joints. Part number CGB-125-09 is used as a replacement neck for 2L Squibb style separatory funnels.
Catalog No. 50-153-1916 Supplier Chemglass Life Sciences Supplier No. CGB12501
#9 Outer Joint, 11mm X 1.5W Tubing. Standard taper flask length outer joints. Part number CGB-125-09 is used as a replacement neck for 2L Squibb style separatory funnels.