Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 9-Tricosene, (9Z)- | 27519-02-4 | 98.0% | 322.61 | 500 MG
SDP

Catalog No. 5000453374
Encompass_Preferred

(Z)-9-Tricosene (cis-9-Tricosene) is an insect pheromone found in flies of the order Diptera, such as the housefly, and can be utilized as an insecticide. It is intended for research use only.

  • Appearance: Colorless to light yellow liquid
  • Initial source: Animals (Musca domestica)
  • Storage (pure form): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month
  • Solubility (in vitro): 10 mg/mL in DMSO (requires ultrasonic, warming, and heating to 60°C)

Catalog No. 50-004-53374 Supplier Medchemexpress LLC Supplier No. HYW009165500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Z)-9-Tricosene (cis-9-Tricosene) is an insect pheromone found in flies of the order Diptera, such as the housefly, and can be utilized as an insecticide. It is intended for research use only.

  • Appearance: Colorless to light yellow liquid
  • Initial source: Animals (Musca domestica)
  • Storage (pure form): -20°C for 3 years, 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month
  • Solubility (in vitro): 10 mg/mL in DMSO (requires ultrasonic, warming, and heating to 60°C)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.