Learn More
Accuflow Systems Saint-Gobain Gas Sampling Bag, PVF, 9 x 9 in, On/Off
Catalog No. NC3245382
Encompass_Preferred
Reusable 9 x 9 in gas sampling bag constructed from 2 mil PVF (Tedlar) fluoropolymer film with heat-sealed seams and an on/off valve for secure collection and temporary storage of noncorrosive gas samples in laboratory environments.
  • Constructed from 2 mil PVF (Tedlar) fluoropolymer film
  • Dimensions 9 x 9 in (230 x 230 mm)
  • Nickel-plated brass on/off valve accepts 1/4 in I.D. tubing or PTFE/silicone septum in polypropylene housing
  • Heat-sealed seams for leak-resistant operation
  • Low permeability to limit sample loss during storage
  • Individually packaged, sold in packs of 10
  • Suitable for sampling noncorrosive gases such as CO, SO2, H2S, radon, and mercaptans
  • Referenced for use in Title 40 CFR Part 60, Method 18 applications where applicable
Catalog No. NC3245382 Supplier Accuflow Systems Supplier No. D107501010
Reusable 9 x 9 in gas sampling bag constructed from 2 mil PVF (Tedlar) fluoropolymer film with heat-sealed seams and an on/off valve for secure collection and temporary storage of noncorrosive gas samples in laboratory environments.
  • Constructed from 2 mil PVF (Tedlar) fluoropolymer film
  • Dimensions 9 x 9 in (230 x 230 mm)
  • Nickel-plated brass on/off valve accepts 1/4 in I.D. tubing or PTFE/silicone septum in polypropylene housing
  • Heat-sealed seams for leak-resistant operation
  • Low permeability to limit sample loss during storage
  • Individually packaged, sold in packs of 10
  • Suitable for sampling noncorrosive gases such as CO, SO2, H2S, radon, and mercaptans
  • Referenced for use in Title 40 CFR Part 60, Method 18 applications where applicable