Learn More
Cayman Chemical 9 Z 11 E 13 EOctadecatrienoic
SDP
Catalog No. 502887708
Encompass_Preferred
α-ESA methyl ester is a neutral, more lipid soluble form of the free acid.
Catalog No. 50-288-7708 Supplier Cayman Chemical Supplier No. 100083331 MG
α-ESA methyl ester is a neutral, more lipid soluble form of the free acid.