Learn More
Cayman Chemical 9 Z 11 E 13 EOctadecatrienoic
SDP
Supplier:  Cayman Chemical 100083495 MG
Encompass_Preferred
A fatty acid agonist of PPARγ; activates PPARγ transcriptional activity in a reporter assay using RAW 264.7 cells at 10 µM; an inhibitor of DNA polymerase α, -β, -γ, -Δ, and -ε (IC50s = 10.7, 13.4, 10.6, 14, and 22 µM, respectively); an inhibitor of topoisomerase I and -II (IC50s = 20 and 5 µM, respectively); reduces the viability of, and induces apoptosis in, MCF-7 breast cancer cells in a concentration-dependent manner; induces ferroptosis in MDA-MB-231 breast cancer cells in an ACSL1-dependent manner; reduces clinical signs of disease in a model of DSS-induced IBD in PPARγ-expressing, but not PPARγ-knockout, mice
Catalog No. 50-288-9295