Learn More
Cayman Chemical 9(Z),11(E),13(Z)-Octadecatrienoic Acid, Purity: ≥95%, CAS: 544-72-9, 10 mg
SDP
Catalog No. 502935691
Encompass_Preferred
An ω-5 long-chain polyunsaturated fatty acid; has been found as a major component in pomegranate seed oil; binds to the AF2 domain of PPARγ (IC50 = 2.5 µM); increases PPARα and PPARγ activity in 3T3-L1 cells in a reporter assay
Catalog No. 50-293-5691 Supplier Cayman Chemical Supplier No. 2605710 MG
An ω-5 long-chain polyunsaturated fatty acid; has been found as a major component in pomegranate seed oil; binds to the AF2 domain of PPARγ (IC50 = 2.5 µM); increases PPARα and PPARγ activity in 3T3-L1 cells in a reporter assay