Learn More
Cayman Chemical 9 Z 11 E 13 ZOctadecatrIe 10mg
SDP
Catalog No. 502933650
Encompass_Preferred
A fatty acid methyl ester; isomer of 9(Z),11(E),13(E)-octadecatrienoic acid methyl ester; has been used as a standard for the quantification of 9(Z),11(E),13(Z)-octadecatrienoic acid in wild growing pomegranate seed oil
Catalog No. 50-293-3650 Supplier Cayman Chemical Supplier No. 2480710 MG
A fatty acid methyl ester; isomer of 9(Z),11(E),13(E)-octadecatrienoic acid methyl ester; has been used as a standard for the quantification of 9(Z),11(E),13(Z)-octadecatrienoic acid in wild growing pomegranate seed oil