Learn More
Cayman Chemical 9 Z 12 Z 15 Z 18 Z 21 ZTetrac
Supplier: Cayman Chemical 10005157250 UG
A very long-chain polyunsaturated fatty acid; has been used to study the desaturation and elongation of ω-3 polyunsaturated fatty acids; found at low levels in mouse brain and spleen and levels decrease in the brain and spleen of old and exceptionally old mice