Learn More
Cayman Chemical 9(Z)-Pentacosene, Purity: ≥95%, CAS: 51865-00-0, 10 mg
SDP
Catalog No. 502892928
Encompass_Preferred
A cuticular hydrocarbon and insect sex hormone; found in the cuticle of female F. canicularis flies and is an attractant to males in a trap assay; a contact sex hormone in scarab beetles and locust borers that is more abundant in females than males; elicits arrestment, alignment, and mounting by males of both species in mating assays, but only elicits copulation by the locust borer; produced by the orchid O. exaltata and by C. cunicularius, a pollinator of O. exaltata
Catalog No. 50-289-2928 Supplier Cayman Chemical Supplier No. 900280710 MG
A cuticular hydrocarbon and insect sex hormone; found in the cuticle of female F. canicularis flies and is an attractant to males in a trap assay; a contact sex hormone in scarab beetles and locust borers that is more abundant in females than males; elicits arrestment, alignment, and mounting by males of both species in mating assays, but only elicits copulation by the locust borer; produced by the orchid O. exaltata and by C. cunicularius, a pollinator of O. exaltata