Learn More
Cayman Chemical 9 Z-TrIcosen 1g
SDP
Catalog No. 502952170
Encompass_Preferred
An insect pheromone used in pesticides; activates antennal basiconic Or7a receptors in Drosophila; produced by honey bees to communicate information about food sources
Catalog No. 50-295-2170 Supplier Cayman Chemical Supplier No. 132361 G
An insect pheromone used in pesticides; activates antennal basiconic Or7a receptors in Drosophila; produced by honey bees to communicate information about food sources