Learn More
Grainger 90 Ballcock Elbow 1/2 CTSx7/8-15/16 UNS
Supplier:  Grainger 16T755
Encompass_Preferred
The John Guest Female Ballcock Elbow is a plastic elbow fitting for use in toilet tank ballcocks. It has a 1/2" female NPT thread on one end to connect to the ballcock fill valve and a 3/8" compression fitting on the other end to connect to the toilet tank refill tube. The elbow allows adjusting the angle of the refill tube. It is made from plastic for corrosion resistance. This elbow provides a reliable connection method for directing water from the ballcock to refill the toilet tank after flushing.
Catalog No. 50-276-8619