Learn More
Grainger 90 Elbow, 316 Stainless Steel
Catalog No. NC2592578
Encompass_Preferred
The 90° Elbow 316 Stainless Steel Pipe Fitting is made from 316 stainless steel for corrosion resistance. It has a 90 degree angle and is designed for use in process piping applications. This elbow fitting has beveled ends that allow for easy welding. It meets ASME B16.9 specifications and is rated for pressures up to 4,000 psi at 650°F. The fitting has a wall thickness of 0.154 inches and is available in pipe sizes ranging from 1/2 inch to 4 inches. This elbow provides a smooth 90 degree turn in process piping systems.
Catalog No. NC2592578 Supplier Grainger Supplier No. 11D020
The 90° Elbow 316 Stainless Steel Pipe Fitting is made from 316 stainless steel for corrosion resistance. It has a 90 degree angle and is designed for use in process piping applications. This elbow fitting has beveled ends that allow for easy welding. It meets ASME B16.9 specifications and is rated for pressures up to 4,000 psi at 650°F. The fitting has a wall thickness of 0.154 inches and is available in pipe sizes ranging from 1/2 inch to 4 inches. This elbow provides a smooth 90 degree turn in process piping systems.