Learn More
Grainger 90 Drop Ear Elbow 1/2 NPT Brass Wht
Catalog No. 502767259
Encompass_Preferred
The Grainger 16T774 is a 16-ton hydraulic bottle jack made by Hein-Werner. It has a lifting range of 10-3/8 inches to 16-1/4 inches and a lowered height of 9-3/8 inches. This compact, portable jack has a welded base for stability and swivel saddle to adjust to any angle. It is designed for use on hard level surfaces and has a built-in safety valve and bypass device. This heavy duty hydraulic jack has a weight capacity of 32,000 lbs.
Catalog No. 50-276-7259 Supplier Grainger Supplier No. 16T774
The Grainger 16T774 is a 16-ton hydraulic bottle jack made by Hein-Werner. It has a lifting range of 10-3/8 inches to 16-1/4 inches and a lowered height of 9-3/8 inches. This compact, portable jack has a welded base for stability and swivel saddle to adjust to any angle. It is designed for use on hard level surfaces and has a built-in safety valve and bypass device. This heavy duty hydraulic jack has a weight capacity of 32,000 lbs.