Learn More
Grainger 90 Elbow Forged Steel 1 in Female NPT
Catalog No. 502750635
Encompass_Preferred
The Grainger 14J731 is a 14-inch diameter, 731 CFM, direct drive, aluminum exhaust fan. It features a 1/3 HP, 115V motor with thermal overload protection and sealed ball bearings for continuous duty. The fan has a galvanized steel housing with baked enamel finish and comes with wall mounting brackets. It is suitable for commercial and industrial exhaust ventilation applications. The maximum operating temperature is 104°F and it weighs approximately 26 lbs.
Catalog No. 50-275-0635 Supplier Grainger Supplier No. 14J731
The Grainger 14J731 is a 14-inch diameter, 731 CFM, direct drive, aluminum exhaust fan. It features a 1/3 HP, 115V motor with thermal overload protection and sealed ball bearings for continuous duty. The fan has a galvanized steel housing with baked enamel finish and comes with wall mounting brackets. It is suitable for commercial and industrial exhaust ventilation applications. The maximum operating temperature is 104°F and it weighs approximately 26 lbs.