Learn More
Grainger 90 Elbow Forged Steel 2 in Female NPT
Catalog No. 502751028
Encompass_Preferred
The Anvil 90 Degree Elbow Forged Steel Pipe Fitting is made from forged steel for durability. It has a 90 degree elbow shape to allow two pipes to connect at a right angle. The fitting has female National Pipe Thread ends to connect to threaded pipe. It is rated for a maximum pressure of 2000 psi at 100 degrees Fahrenheit. The elbow has a black finish and is 3 inches in size. This elbow fitting allows you to route pipe around corners in hydraulic, steam, gas and air systems.
Catalog No. 50-275-1028 Supplier Grainger Supplier No. 14J733
The Anvil 90 Degree Elbow Forged Steel Pipe Fitting is made from forged steel for durability. It has a 90 degree elbow shape to allow two pipes to connect at a right angle. The fitting has female National Pipe Thread ends to connect to threaded pipe. It is rated for a maximum pressure of 2000 psi at 100 degrees Fahrenheit. The elbow has a black finish and is 3 inches in size. This elbow fitting allows you to route pipe around corners in hydraulic, steam, gas and air systems.