Learn More
Grainger 90 Elbow Forged Steel 3/4 in FNPT
Catalog No. 502751027
Encompass_Preferred
The Anvil 90 Degree Elbow Forged Steel Pipe Fitting is made from forged steel for durability. It has a 90 degree elbow shape to change pipe direction and is rated for 150 psi working pressure at up to 650 degrees Fahrenheit. This 1/2 inch NPT threaded elbow fitting is manufactured in the USA and is commonly used in steam, water, oil, natural gas and air applications. The elbow has a black finish to resist corrosion.
Catalog No. 50-275-1027 Supplier Grainger Supplier No. 14J730
The Anvil 90 Degree Elbow Forged Steel Pipe Fitting is made from forged steel for durability. It has a 90 degree elbow shape to change pipe direction and is rated for 150 psi working pressure at up to 650 degrees Fahrenheit. This 1/2 inch NPT threaded elbow fitting is manufactured in the USA and is commonly used in steam, water, oil, natural gas and air applications. The elbow has a black finish to resist corrosion.