Learn More
Grainger 90 Elbow SS 2 in Socket x Push Fit
Catalog No. 502752429
Encompass_Preferred
The Blucher 90° Sweep Elbow is a stainless steel elbow fitting with a 90 degree sweep. It has a 2 inch nominal pipe size and is made from 304 stainless steel for corrosion resistance. The elbow has a smooth interior surface and is welded construction. It can withstand pressures up to 300 psi at 70°F. The elbow is ideal for use in sanitary drainage, waste, and vent piping applications.
Catalog No. 50-275-2429 Supplier Grainger Supplier No. 14N968
The Blucher 90° Sweep Elbow is a stainless steel elbow fitting with a 90 degree sweep. It has a 2 inch nominal pipe size and is made from 304 stainless steel for corrosion resistance. The elbow has a smooth interior surface and is welded construction. It can withstand pressures up to 300 psi at 70°F. The elbow is ideal for use in sanitary drainage, waste, and vent piping applications.