Learn More
Grainger 90 Elbow Union 1/2 in CTS PEX White
Catalog No. 502764002
Encompass_Preferred
The Grainger 1/2" CTS PEX 90 Degree Elbow Union is a white PEX plastic fitting used to connect PEX tubing at a 90 degree angle. It has a maximum pressure rating of 100 PSI at 73°F and is NSF-pw certified for use with potable water. The elbow union has 1/2" CTS connections on both ends and is compatible with PEX tubing. This elbow fitting allows for easy installation and creates secure joints without requiring clamps or crimp rings. It can be used for hot and cold potable water applications in residential plumbing systems. The elbow is lead-free and made in the USA.
Catalog No. 50-276-4002 Supplier Grainger Supplier No. 16T734
The Grainger 1/2" CTS PEX 90 Degree Elbow Union is a white PEX plastic fitting used to connect PEX tubing at a 90 degree angle. It has a maximum pressure rating of 100 PSI at 73°F and is NSF-pw certified for use with potable water. The elbow union has 1/2" CTS connections on both ends and is compatible with PEX tubing. This elbow fitting allows for easy installation and creates secure joints without requiring clamps or crimp rings. It can be used for hot and cold potable water applications in residential plumbing systems. The elbow is lead-free and made in the USA.