Learn More
Grainger 90 Elbow Union 1 in CTS PEX White
Catalog No. 502768437
Encompass_Preferred
The 1-inch 90 degree PEX elbow union by Apollo is a white plastic fitting used to connect PEX tubing at a 90 degree angle. It has a maximum temperature rating of 180°F and maximum pressure rating of 160 PSI. The fitting has a CTS (copper tube size) on one end and is compatible with 1-inch PEX tubing on the other end. This elbow fitting allows for convenient 90 degree turns when installing PEX plumbing lines. It provides a secure connection and is made from durable PEX plastic that is corrosion and scale resistant.
Catalog No. 50-276-8437 Supplier Grainger Supplier No. 16T736
The 1-inch 90 degree PEX elbow union by Apollo is a white plastic fitting used to connect PEX tubing at a 90 degree angle. It has a maximum temperature rating of 180°F and maximum pressure rating of 160 PSI. The fitting has a CTS (copper tube size) on one end and is compatible with 1-inch PEX tubing on the other end. This elbow fitting allows for convenient 90 degree turns when installing PEX plumbing lines. It provides a secure connection and is made from durable PEX plastic that is corrosion and scale resistant.