Learn More
Grainger 90 Stem Elbow 3/4 x 3/4 Tube PEX White
Catalog No. 502768617
Encompass_Preferred
The Grainger Approved 90° Stem Elbow is a PVC fitting with a 90 degree angle. It has a socket end and a spigot end to connect PVC pipes. The elbow has a maximum temperature rating of 140°F and is made of PVC plastic for corrosion resistance. This elbow fitting allows you to change direction in a PVC pipe system. It is designed for use in industrial plumbing and process piping applications. The elbow has a pressure rating of up to 280 psi and comes in sizes ranging from 1/2" to 4". It meets ASTM D2466 standards.
Catalog No. 50-276-8617 Supplier Grainger Supplier No. 16T746
The Grainger Approved 90° Stem Elbow is a PVC fitting with a 90 degree angle. It has a socket end and a spigot end to connect PVC pipes. The elbow has a maximum temperature rating of 140°F and is made of PVC plastic for corrosion resistance. This elbow fitting allows you to change direction in a PVC pipe system. It is designed for use in industrial plumbing and process piping applications. The elbow has a pressure rating of up to 280 psi and comes in sizes ranging from 1/2" to 4". It meets ASTM D2466 standards.