Learn More
Grainger 90 Sweep Elbow SS 2 Socket x Push Fit
Catalog No. 502748899
Encompass_Preferred
The Blucher 90 Sweep Elbow is a stainless steel elbow fitting with a 90 degree sweep. It has a 2 inch diameter and is made from 304 stainless steel for corrosion resistance. The elbow has a smooth interior surface and is welded for maximum strength. It can withstand temperatures up to 850 degrees Fahrenheit. The elbow fitting allows a 90 degree change of direction in 2 inch stainless steel tubing runs. It is commonly used in food, dairy, pharmaceutical, and chemical processing applications. The elbow has a pressure rating of 285 psi and conforms to ASTM A270 standards. It enables efficient flow while minimizing turbulence.
Catalog No. 50-274-8899 Supplier Grainger Supplier No. 14N971
The Blucher 90 Sweep Elbow is a stainless steel elbow fitting with a 90 degree sweep. It has a 2 inch diameter and is made from 304 stainless steel for corrosion resistance. The elbow has a smooth interior surface and is welded for maximum strength. It can withstand temperatures up to 850 degrees Fahrenheit. The elbow fitting allows a 90 degree change of direction in 2 inch stainless steel tubing runs. It is commonly used in food, dairy, pharmaceutical, and chemical processing applications. The elbow has a pressure rating of 285 psi and conforms to ASTM A270 standards. It enables efficient flow while minimizing turbulence.