Learn More
Grainger 90 Swivel Elbow 1/2 CTSx1/2 NPS PEX
Catalog No. 502765086
Encompass_Preferred
The Grainger 16T754 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA standard 56C frame with a 1.15 service factor. The motor is totally enclosed and fan cooled with Class F insulation and a C-face endshield for easy mounting. It can provide 0.56 kW of output power. The 16T754 is built for general purpose applications requiring up to 3/4 horsepower.
Catalog No. 50-276-5086 Supplier Grainger Supplier No. 16T754
The Grainger 16T754 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA standard 56C frame with a 1.15 service factor. The motor is totally enclosed and fan cooled with Class F insulation and a C-face endshield for easy mounting. It can provide 0.56 kW of output power. The 16T754 is built for general purpose applications requiring up to 3/4 horsepower.