Learn More
VEOLIA WTS 900 1/MG CAL 500 AEG/L VER SET
Catalog No. NC0552099
Encompass
Single point calibration and verification standards set for Sievers 900 Series. Includes 1 TOC Calibration Set (1 mg C/L) and 1 Verification Set (500 ug C/L)
Catalog No. NC0552099 Supplier VEOLIA WTS Supplier No. STD 90026
Single point calibration and verification standards set for Sievers 900 Series. Includes 1 TOC Calibration Set (1 mg C/L) and 1 Verification Set (500 ug C/L)