Learn More
Electron Microscopy Sciences Grinding and Polishing Machine Model 900 CE MARK
SDP
Catalog No. 50980887
Encompass
This light weight, bench top grinding/polishing model is ideal for surface preparation of small metallographic specimens. It is adjustable in speed from 0 to 1725 RPM, and has 1/3 HP and it is fully grounded. The coolant is applied to the wheel through an adjustable flow control goose-neck spout. The unit comes complete with a removable rubber splash guard for cleaning and a compression fitting for an easy connection into the existing coolant supply.
Model 900 includes aluminum lapping plate, lapping plate removal wrench, metal splash guard, metal holding band (for plain back paper) and rubber splash guard. CE MARK.
Catalog No. 50-980-887 Supplier Electron Microscopy Sciences Supplier No. 5012020
This light weight, bench top grinding/polishing model is ideal for surface preparation of small metallographic specimens. It is adjustable in speed from 0 to 1725 RPM, and has 1/3 HP and it is fully grounded. The coolant is applied to the wheel through an adjustable flow control goose-neck spout. The unit comes complete with a removable rubber splash guard for cleaning and a compression fitting for an easy connection into the existing coolant supply.
Model 900 includes aluminum lapping plate, lapping plate removal wrench, metal splash guard, metal holding band (for plain back paper) and rubber splash guard. CE MARK.