Learn More
VEOLIA WTS 900 SINGLE POINT VERIFICATION
Catalog No. NC0397470
Encompass
Single point verification standards set for Sievers 900 Series. Includes: one Verification Blank, one 25 mg C/L TOC Verification Standard (as NIST Sucrose) and one 25 mg C/L IC Standard (as Na2CO3).
Catalog No. NC0397470 Supplier VEOLIA WTS Supplier No. STD 9001201
Single point verification standards set for Sievers 900 Series. Includes: one Verification Blank, one 25 mg C/L TOC Verification Standard (as NIST Sucrose) and one 25 mg C/L IC Standard (as Na2CO3).