Learn More
Sino Biological Recombinant Cynomolgus, Rhesus TNF-alpha Protein
Supplier:  Sino Biological 90018CNAE20
Encompass_Preferred
A DNA sequence encoding the cynomolgus / rhesus TNF [(Identical to the rhesus TNF (NP_001040614.1)] (Val77-Leu233) was expressed and purified with an initial Met. Cynomolgus and Rhesus TNF sequences are identical.
Catalog No. 50-202-1659