Learn More
Sino Biological Recombinant Cynomolgus, Rhesus M-CSF/CSF1 Protein (hFc Tag)
Supplier:  Sino Biological 90029C02H50
Encompass_Preferred
A DNA sequence encoding the cynomolgus / rhesus CSF1 (XP_005542489.1) (Met1-Asn190) was expressed with the Fc region of human IgG1 at the C-terminus. Cynomolgus and Rhesus CSF1 sequences are identical.
Catalog No. 50-202-1682