Learn More
Sino Biological Recombinant Rhesus HER3/ERBB3 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 90043K08H100
Encompass_Preferred
A DNA sequence encoding the rhesus ErbB3 (XP_001113953.2) extracellular domain (Met 1-Thr 643) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-202-1715