Learn More
Hach Company 900Max 4-20mA Interface
Supplier:  Hach Company 2021
Encompass
4-20mA interface-10PPM FS-Out Converts analog signal from flow meter to pulses compatible with 900Max sampler
Catalog No. 50-002-56975