Learn More
Quality Lab Accessories 900ML CLR RND VOL FLASK
Catalog No. NC9428482
Encompass
900mL Volumetric Flask with Round Bottom, Class A, Calibrated to 20�C
Catalog No. NC9428482 Supplier Quality Lab Accessories Supplier No. VOLFLK09
900mL Volumetric Flask with Round Bottom, Class A, Calibrated to 20�C