Learn More
Ace Glass, Inc. Flask, freeze drying, 900mL, 199mm height, 53mm ID neck, 100mm body OD, includes snap-on rubber cap and 1/2in ODPTFE plain manifold connection adapter
SDP
Catalog No. 5097426
Encompass_Preferred
Catalog No. 50-974-26 Supplier Ace Glass, Inc. Supplier No. 703521