Learn More
Quality Lab Accessories 900ML VOL. FLK CLS A CAL.37C
Catalog No. NC9022417
Encompass
900mL Volumetric Flask with Round Bottom, Class A, Calibrated to 37�C
Catalog No. NC9022417 Supplier Quality Lab Accessories Supplier No. VOLFLKX9
900mL Volumetric Flask with Round Bottom, Class A, Calibrated to 37�C