Learn More
Sino Biological Recombinant Cynomolgus, Rhesus TNFR-2 / CD120b Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 90102C08H50
Encompass_Preferred
A DNA sequence encoding the cynomolgus / rhesus TNFRSF1B (XP_005544817.1 & NP_001253134.1) (Met1-Asp257) was expressed, fused with a polyhistidine tag at the C-terminus. Cynomolgus and Rhesus TNFRSF1B sequences are identical.
Catalog No. 50-202-1820