Learn More
Sino Biological Recombinant Cynomolgus IL-18Rα Protein (hFc Tag)
Supplier:  Sino Biological 90121C02H100
Encompass_Preferred
A DNA sequence encoding the cynomolgus IL18R1 (F7H8F9) (Met1-Lys315) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-202-1870